This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GM9573 SIRNA Assays ›
Gene Symbol:
Gm9573
Gene Name:
predicted gene 9573
Gene Aliases:
Muc21
Chromosome Location:
Chr.17: 35617923 - 35626637 on Build GRCm38
UniGene ID:
Mm.454025
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s127235

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gm9573
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244654.1 NP_001231583.1 1718
GenBank mRNA HM596785.1 - 1720

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell-substrate adhesion
negative regulation of cell-cell adhesion
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0