This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4930467E23RIK SIRNA Assays ›
Gene Symbol:
4930467E23Rik Gm15319 Gm21119
Gene Name:
RIKEN cDNA 4930467E23 gene
predicted gene 15319
predicted gene, 21119
Gene Aliases:
ENSMUSG00000074453, OTTMUSG00000018948, TSAP
OTTMUSG00000020118
-
Chromosome Location:
Chr.8: 19729576 - 19753603 on Build GRCm38
Chromosome Location:
Chr.8: 20337664 - 20363202 on Build GRCm38
Chromosome Location:
Chr.8: 20614405 - 20657119 on Build GRCm38
UniGene ID:
Mm.425517
Mm.491087
Mm.491087
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s126734

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930467E23Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039553.2 NP_001034642.2 1 375
Gene Symbol: Gm15319
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177408.1 NP_001170879.1 1 375
GenBank mRNA BC150794.1 - 1 227 9087705
Gene Symbol: Gm21119
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270553.1 NP_001257482.1 1 375
GenBank mRNA AK076593.1 - 1 376

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0