siRNA Details
Gene Symbol: Gm21119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270553.1 | NP_001257482.1 | 5 | 864 | ||
GenBank mRNA | AK076593.1 | - | 5 | 865 |
Gene Symbol: 4930467E23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039553.2 | NP_001034642.2 | 5 | 864 |
Gene Symbol: Gm15319 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177408.1 | NP_001170879.1 | 5 | 864 | ||
GenBank mRNA | BC150794.1 | - | 5 | 716 | 9087705 |
Gene Symbol: Gm3643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029875.1 | - | 1603 |
