This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIEZO2 SIRNA Assays ›
Gene Symbol:
Piezo2
Gene Name:
piezo-type mechanosensitive ion channel component 2
Gene Aliases:
5930434P17, 9030411M15Rik, 9430028L06Rik, Fam38b, Fam38b2
Chromosome Location:
Chr.18: 63010213 - 63387307 on Build GRCm38
UniGene ID:
Mm.158720
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s125987

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Piezo2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039485.4 NP_001034574.4 25 4574
GenBank mRNA AK050232.1 - 17 2881
AK087529.1 - 24 3805
HQ215521.1 - 25 3660

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
response to mechanical stimulus
regulation of membrane potential
response to stimulus
detection of mechanical stimulus involved in sensory perception
cation transmembrane transport
cation channel activity
protein binding
mechanically-gated ion channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0