This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BC080696 SIRNA Assays ›
Gene Symbol:
BC080696 A430089I19Rik
Gene Name:
cDNA sequence BC080696
RIKEN cDNA A430089I19 gene
Gene Aliases:
-
Gm16522
Chromosome Location:
Chr.5: 94434131 - 94436940 on Build GRCm38
Chromosome Location:
Chr.5: 94302519 - 94306796 on Build GRCm38
UniGene ID:
Mm.380154
Mm.26025
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s125394

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BC080696
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC080696.1 - 3 1763
Gene Symbol: A430089I19Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177913.4 NP_808581.1 4 1838
GenBank mRNA AK040373.1 - 4 1839

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm