This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCML1 SIRNA Assays ›
Gene Symbol:
SCML1
Gene Name:
sex comb on midleg-like 1 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.X: 17737449 - 17754988 on Build GRCh38
UniGene ID:
Hs.109655
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s12509

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SCML1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037535.2 NP_001032624.1 4 693
NM_001037536.2 NP_001032625.1 4 690
NM_001037540.2 NP_001032629.1 6 923
NM_006746.5 NP_006737.2 5 842
XM_005274578.2 XP_005274635.1 6 918
XM_005274579.2 XP_005274636.1 5 831
XM_006724508.3 XP_006724571.1 5 3823
XM_006724509.3 XP_006724572.1 4 3739
XM_011545564.2 XP_011543866.1 5 3826
XM_017029723.1 XP_016885212.1 4 3742
GenBank mRNA AF160728.1 - 4 630
AK313993.1 - 4 651
BC026159.2 - 5 822 4827364
BC105028.1 - 2 299 8144031
BX647757.1 - 3 2607

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
anatomical structure morphogenesis
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0