This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TCTN1 SIRNA Assays ›
Gene Symbol:
Tctn1
Gene Name:
tectonic family member 1
Gene Aliases:
G730031O11Rik, Tect1
Chromosome Location:
Chr.5: 122239495 - 122264489 on Build GRCm38
UniGene ID:
Mm.28804
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s124825

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tctn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039153.3 NP_001034242.2 3 566
GenBank mRNA AK036568.1 - 3 548
AK138661.1 - 3 565
AK144772.1 - 3 542
DQ278867.1 - 3 567

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
neural tube formation
multicellular organism development
regulation of smoothened signaling pathway
somatic motor neuron differentiation
telencephalon development
dorsal/ventral neural tube patterning
central nervous system interneuron axonogenesis
cell projection organization
cilium morphogenesis
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0