This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TSPAN31 SIRNA Assays ›
Gene Symbol:
TSPAN31
Gene Name:
tetraspanin 31
Gene Aliases:
SAS
Chromosome Location:
Chr.12: 57744090 - 57748243 on Build GRCh38
UniGene ID:
Hs.632708
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s12474

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TSPAN31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005981.3 NP_005972.1 6 743
XM_005269074.3 XP_005269131.2 6 1668
XM_017019793.1 XP_016875282.1 4 472
GenBank mRNA AK090502.1 - 4 1146
AK294128.1 - 6 696
AK315765.1 - 6 864
BC010377.1 - 6 696 4294627
BX647402.1 - 4 469
CR407647.1 - 6 589
U01160.1 - 6 691

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cell surface receptor signaling pathway
positive regulation of cell proliferation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0