This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TDPOZ4 SIRNA Assays ›
Gene Symbol:
Tdpoz4 Gm4858
Gene Name:
TD and POZ domain containing 4
predicted gene 4858
Gene Aliases:
-
EG229571
Chromosome Location:
Chr.3: 93796398 - 93797510 on Build GRCm38
Chromosome Location:
Chr.3: 93068823 - 93075505 on Build GRCm38
UniGene ID:
Mm.377924
Mm.331194
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s119490

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tdpoz4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207272.2 NP_997155.2 1 275
GenBank mRNA BC115626.1 - 1 275 40102845
BC115627.1 - 1 271
Gene Symbol: Gm4858
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177694.1 NP_001171165.1 1 275
GenBank mRNA BC172161.1 - 1 259

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0