This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF4A1 SIRNA Assays ›
Gene Symbol:
Eif4a1
Gene Name:
eukaryotic translation initiation factor 4A1
Gene Aliases:
BM-010, Ddx2a, Eif4
Chromosome Location:
Chr.11: 69666936 - 69672423 on Build GRCm38
UniGene ID:
Mm.26680
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s119380

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif4a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159375.1 NP_001152847.1 6, 7 755
NM_144958.4 NP_659207.1 6, 7 755
GenBank mRNA AK010644.1 - 6, 7 657
AK011529.1 - 6, 7 648
AK077429.1 - 6, 7 648
AK088124.1 - 4 1973
AK135752.1 - 6, 7 648
AK135994.1 - 6, 7 649
AK144850.1 - 6, 7 648
AK145375.1 - 6, 7 650
AK145508.1 - 6, 7 650
AK146424.1 - 6, 7 650
AK146464.1 - 6, 7 648
AK152210.1 - 3, 4 431
AK152694.1 - 6, 7 650
AK159643.1 - 6, 7 650
AK162017.1 - 650
AK166439.1 - 6, 7 648
AK166527.1 - 6, 7 648
AK166572.1 - 6, 7 650
AK166674.1 - 6, 7 650
AK167003.1 - 6, 7 648
AK167083.1 - 6, 7 648
AK167196.1 - 6, 7 650
AK167205.1 - 6, 7 647
AK167906.1 - 6, 7 650
AK167988.1 - 6, 7 648
AK168697.1 - 6, 7 649
AK168780.1 - 6, 7 648
AK169023.1 - 6, 7 648
AK169030.1 - 6, 7 641
AK169057.1 - 6, 7 648
AK169156.1 - 6, 7 648
AK169279.1 - 6, 7 648
AK185730.1 - 2, 3 132
AK187979.1 - 1, 2 132
AK196145.1 - 132
AK204159.1 - 2, 3 295
AK213871.1 - 3, 4 306
AK215233.1 - 1, 2 132
AK219842.1 - 1, 2 132
BC012282.1 - 6, 7 637
BC049915.1 - 6, 7 638 5068709
BC099392.1 - 5, 6 631
X03039.1 - 6, 7 645
X03040.1 - 5, 6 648

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of gene expression
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
double-stranded RNA binding
translation initiation factor activity
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0