This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THOC6 SIRNA Assays ›
Gene Symbol:
Thoc6
Gene Name:
THO complex 6 homolog (Drosophila)
Gene Aliases:
F830014G06Rik, Wdr58
Chromosome Location:
Chr.17: 23668618 - 23673903 on Build GRCm38
UniGene ID:
Mm.328831
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s118572

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thoc6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008425.1 NP_001008425.1 9 840
GenBank mRNA AK089746.1 - 6 1168
BC085131.1 - 9 840 5715435
BC089551.1 - 10 840 30303151

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_024384, AF275266, BC079149.

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
mRNA export from nucleus
transport
apoptotic process
multicellular organism development
central nervous system development
RNA splicing
negative regulation of apoptotic process
viral mRNA export from host cell nucleus
mRNA transport
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0