This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF133 SIRNA Assays ›
Gene Symbol:
Rnf133
Gene Name:
ring finger protein 133
Gene Aliases:
Greul2
Chromosome Location:
Chr.6: 23648869 - 23650305 on Build GRCm38
UniGene ID:
Mm.436547
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s118568

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf133
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198251.2 NP_937894.1 1 630
GenBank mRNA AK028082.1 - 1 631
AY155441.1 - 1 259
BC116423.1 - 1 531 40103909
BC116424.1 - 1 531 40103912

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein autoubiquitination
molecular_function
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0