This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YPEL5 SIRNA Assays ›
Gene Symbol:
Ypel5
Gene Name:
yippee-like 5 (Drosophila)
Gene Aliases:
2310076K21Rik, CGI-127
Chromosome Location:
Chr.17: 72836625 - 72851195 on Build GRCm38
UniGene ID:
Mm.277533
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s118249

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ypel5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027166.5 NP_081442.1 3 339
GenBank mRNA AB098744.1 - 2 227
AK010201.1 - 3 305
AK046281.1 - 3 338
AK049096.1 - 3 340
AK049644.1 - 3 338
AK137446.1 - 3 311
AK154461.1 - 3 378
AK165943.1 - 3 341
AK167998.1 - 3 355
AK169241.1 - 3 342
AK169986.1 - 3 313
AK171010.1 - 3 340
BC025125.1 - 1 99 4921751
BC085109.1 - 3 358 30737658

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001127399, NM_016061, NM_001127401, NM_001127400, BC047237, and others.
This siRNA also targets the following transcripts in Rat: NM_001035221, XM_008764509, XM_008764508, XM_008764510, CB584328, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0