This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAPSN SIRNA Assays ›
Gene Symbol:
RAPSN
Gene Name:
receptor associated protein of the synapse
Gene Aliases:
CMS11, CMS4C, FADS, RAPSYN, RNF205
Chromosome Location:
Chr.11: 47437757 - 47449178 on Build GRCh38
UniGene ID:
Hs.81218
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s11798

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RAPSN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005055.4 NP_005046.2 7 1322
NM_032645.4 NP_116034.2 5 1145
XM_005253042.3 XP_005253099.1 6 1226
XM_005253043.3 XP_005253100.1 6 1157
XM_011520252.1 XP_011518554.1 7 1365
GenBank mRNA AF449218.1 - 7 1322
AK310002.1 - 5 1103
BC004196.2 - 5 1110 3530551
KU178300.1 - 5 931
KU178301.1 - 7 1244
KU178302.1 - 7 1108
KU178303.1 - 6 1054
KU178304.1 - 6 985
Z33905.1 - 7 1315

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chemical synaptic transmission
synaptic transmission, cholinergic
positive regulation of neuron apoptotic process
skeletal muscle acetylcholine-gated channel clustering
positive regulation of neuromuscular synaptic transmission
regulation of postsynaptic membrane organization
zinc ion binding
acetylcholine receptor binding
ionotropic glutamate receptor binding
protein anchor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0