This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other N4BP2L2 SIRNA Assays ›
Gene Symbol:
N4bp2l2
Gene Name:
NEDD4 binding protein 2-like 2
Gene Aliases:
2700092H06Rik, zag1
Chromosome Location:
Chr.5: 150635973 - 150665612 on Build GRCm38
UniGene ID:
Mm.246934
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s117828

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: N4bp2l2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201369.3 NP_958757.2 2 369
GenBank mRNA AK038740.1 - 2 289
BC037393.1 - 2 334
CF724618.1 - 2 276
CJ162163.1 - 2 370

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001005533, XM_006248735, XM_006248736, BC081998.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
blastocyst development
transcription from RNA polymerase II promoter
positive regulation of hematopoietic stem cell proliferation
negative regulation of hematopoietic stem cell differentiation
RNA polymerase II transcription corepressor activity
protein binding
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0