This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PARL SIRNA Assays ›
Gene Symbol:
Parl
Gene Name:
presenilin associated, rhomboid-like
Gene Aliases:
D16Ertd607e, PRO2207, PSARL1, PSENIP2, Psarl
Chromosome Location:
Chr.16: 20279820 - 20302395 on Build GRCm38
UniGene ID:
Mm.371737
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s117460

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Parl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005767.4 NP_001005767.1 9, 10 1062
GenBank mRNA AK150980.1 - 9, 10 1068
AK151652.1 - 9, 10 1061
BC083153.1 - 9, 10 1062 30728006

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001035249, BC088226, FQ214115, BC158564, BC105907, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein processing
regulation of proteolysis
negative regulation of release of cytochrome c from mitochondria
regulation of mitophagy
regulation of protein targeting to mitochondrion
regulation of reactive oxygen species metabolic process
negative regulation of intrinsic apoptotic signaling pathway
endopeptidase activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0