This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STMND1 SIRNA Assays ›
Gene Symbol:
Stmnd1
Gene Name:
stathmin domain containing 1
Gene Aliases:
Gm1574
Chromosome Location:
Chr.13: 46273721 - 46300115 on Build GRCm38
UniGene ID:
Mm.188108
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s117388

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stmnd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005422.1 NP_001005422.1 4 575
GenBank mRNA BC064117.1 - 4 575 3418830

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008771561, XM_006253772.

Gene Ontology Categories:

Function(s) Process(es)

microtubule depolymerization
biological_process
regulation of microtubule polymerization or depolymerization
regulation of cytoskeleton organization
molecular_function
tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0