This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NALCN SIRNA Assays ›
Gene Symbol:
Nalcn
Gene Name:
sodium leak channel, non-selective
Gene Aliases:
A530023G15Rik, AI849508, Vgcnl1
Chromosome Location:
Chr.14: 123276634 - 123627363 on Build GRCm38
UniGene ID:
Mm.255348
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s117064

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nalcn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177393.4 NP_796367.3 3 549
GenBank mRNA AK036122.1 - 3 367
AK039034.1 - 3 367
AK040761.1 - 3 541
AK043071.1 - 3 282
AK143219.1 - 3 549

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
ion transmembrane transport
regulation of ion transmembrane transport
transmembrane transport
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
ion channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
cation channel activity
sodium channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0