This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP236 SIRNA Assays ›
Gene Symbol:
Zfp236
Gene Name:
zinc finger protein 236
Gene Aliases:
AI447957, Gm335
Chromosome Location:
Chr.18: 82593593 - 82693627 on Build GRCm38
UniGene ID:
Mm.34438
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s116307

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp236
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177832.3 NP_808500.2 6 742
GenBank mRNA BC138187.1 - 6 742 8861206
BC145179.1 - 6 742
BC145180.1 - 6 742

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008772218, XM_006255016, XM_008772219, XM_006255011, XM_006255013, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0