This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MSLNL SIRNA Assays ›
Gene Symbol:
Mslnl
Gene Name:
mesothelin-like
Gene Aliases:
4732467B22
Chromosome Location:
Chr.17: 25733503 - 25748530 on Build GRCm38
UniGene ID:
Mm.183275
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s116263

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mslnl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177822.3 NP_808490.1 5, 6 686
GenBank mRNA AK028891.1 - 5, 6 686
BC052484.1 - 4, 5 422 6312790

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0