This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM26 SIRNA Assays ›
Gene Symbol:
Tmem26
Gene Name:
transmembrane protein 26
Gene Aliases:
3230401A06, AI195372
Chromosome Location:
Chr.10: 68723746 - 68782655 on Build GRCm38
UniGene ID:
Mm.2479
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s116079

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem26
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177794.3 NP_808462.2 2, 3 421
GenBank mRNA AK028283.1 - 2, 3 422
AK084351.1 - 2, 3 399
AK143866.1 - 2, 3 399
BC117503.1 - 2, 3 343 40106128

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107623.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0