This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MICAL2 SIRNA Assays ›
Gene Symbol:
Mical2
Gene Name:
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases:
5330438E18Rik, 9530064J02, MICAL-2, mKIAA0750
Chromosome Location:
Chr.7: 112225836 - 112355194 on Build GRCm38
UniGene ID:
Mm.488195
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115984

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mical2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193305.1 NP_001180234.1 4 732
NM_177282.5 NP_796256.1 4 732
GenBank mRNA AK030608.1 - 4 713
AK138935.1 - 4 706
AK143191.1 - 4 679
AK161075.1 - 4 679
AK163078.1 - 4 699
AK163231.1 - 4 678
AK220353.1 - 2 409
BC111895.1 - 2 456 40061129
BC112415.1 - 2 456 40061128

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
metabolic process
positive regulation of transcription via serum response element binding
sulfur oxidation
actin filament depolymerization
oxidation-reduction process
actin binding
monooxygenase activity
zinc ion binding
3-(3-hydroxyphenyl)propionate hydroxylase activity
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
4-chlorobenzaldehyde oxidase activity
3,5-xylenol methylhydroxylase activity
phenylacetate hydroxylase activity
4-nitrophenol 4-monooxygenase activity
dimethyl sulfide monooxygenase activity
alpha-pinene monooxygenase [NADH] activity
phenanthrene 9,10-monooxygenase activity
1-hydroxy-2-naphthoate hydroxylase activity
toluene 4-monooxygenase activity
xylene monooxygenase activity
dibenzothiophene monooxygenase activity
6-hydroxy-3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
chlorophenol 4-monooxygenase activity
carbon disulfide oxygenase activity
toluene 2-monooxygenase activity
1-hydroxy-2-oxolimonene 1,2-monooxygenase activity
phenanthrene 1,2-monooxygenase activity
tetrahydrofuran hydroxylase activity
styrene monooxygenase activity
toluene-4-sulfonate monooxygenase activity
toluene-sulfonate methyl-monooxygenase activity
3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
2-hydroxy-phenylacetate hydroxylase activity
2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA 1,2-monooxygenase activity
phenanthrene 3,4-monooxygenase activity
toluene 3-monooxygenase activity
4-hydroxyphenylacetate,NADH:oxygen oxidoreductase (3-hydroxylating) activity
limonene monooxygenase activity
2-methylnaphthalene hydroxylase activity
1-methylnaphthalene hydroxylase activity
bisphenol A hydroxylase A activity
salicylate 5-hydroxylase activity
isobutylamine N-hydroxylase activity
branched-chain dodecylbenzene sulfonate monooxygenase activity
3-HSA hydroxylase activity
4-hydroxypyridine-3-hydroxylase activity
2-octaprenyl-3-methyl-6-methoxy-1,4-benzoquinol hydroxylase activity
6-hydroxynicotinate 3-monooxygenase activity
NADPH:sulfur oxidoreductase activity
metal ion binding
tocotrienol omega-hydroxylase activity
thalianol hydroxylase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0