This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IDI2 SIRNA Assays ›
Gene Symbol:
Idi2
Gene Name:
isopentenyl-diphosphate delta isomerase 2
Gene Aliases:
4833405L16Rik, IPPI2
Chromosome Location:
Chr.13: 8952863 - 8960935 on Build GRCm38
UniGene ID:
Mm.313743
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115796

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Idi2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177197.4 NP_796171.1 2 170
GenBank mRNA AK029364.1 - 2 171
AK081319.1 - 2 171
AY263178.1 - 2 171
BC099586.1 - 2 157 30292324
BC125310.1 - 2 156 40129825
BC138100.1 - 2 156 8861119

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cholesterol biosynthetic process
isopentenyl diphosphate metabolic process
isopentenyl-diphosphate delta-isomerase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0