This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ITPKB SIRNA Assays ›
Gene Symbol:
Itpkb
Gene Name:
inositol 1,4,5-trisphosphate 3-kinase B
Gene Aliases:
1110033J02Rik, E130307H12Rik
Chromosome Location:
Chr.1: 180330476 - 180424802 on Build GRCm38
UniGene ID:
Mm.132535
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115702

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itpkb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081175.1 NP_001074644.1 2 1993
GenBank mRNA AK053759.1 - 2 1942
BC151160.1 - 2 1859 9088049
BC157986.1 - 2 1858 9088142
BC158000.1 - 2 1858 9088148

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_019312, X74227, AJ242781.

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
cell surface receptor signaling pathway
phosphorylation
T cell differentiation
positive thymic T cell selection
thymic T cell selection
positive regulation of Ras protein signal transduction
positive regulation of alpha-beta T cell differentiation
inositol-1,4,5-trisphosphate 3-kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0