This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTPRH SIRNA Assays ›
Gene Symbol:
PTPRH
Gene Name:
protein tyrosine phosphatase, receptor type H
Gene Aliases:
R-PTP-H, SAP1
Chromosome Location:
Chr.19: 55181247 - 55209506 on Build GRCh38
UniGene ID:
Hs.179770
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s11554

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PTPRH
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161440.2 NP_001154912.2 17, 18 2739
NM_002842.4 NP_002833.4 19, 20 3273
XM_011527183.1 XP_011525485.1 19, 20 3294
XM_011527188.1 XP_011525490.1 18, 19 3243
XM_017027056.1 XP_016882545.1 18, 19 3281
XM_017027057.1 XP_016882546.1 18, 19 3269
XM_017027058.1 XP_016882547.1 18, 19 3314
XM_017027060.1 XP_016882549.1 17, 18 3233
XM_017027061.1 XP_016882550.1 16, 17 2747
XM_017027062.1 XP_016882551.1 18, 19 3189
XM_017027063.1 XP_016882552.1 18, 19 3182
GenBank mRNA BC111715.1 - 18, 19 2689 40008777
BC111716.1 - 19, 20 3223 40008775
D15049.2 - 19, 20 3247

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
apoptotic process
peptidyl-tyrosine dephosphorylation
transmembrane receptor protein tyrosine phosphatase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0