This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CELF5 SIRNA Assays ›
Gene Symbol:
Celf5
Gene Name:
CUGBP, Elav-like family member 5
Gene Aliases:
4930565A21Rik, Brunol5
Chromosome Location:
Chr.10: 81459227 - 81482725 on Build GRCm38
UniGene ID:
Mm.152689
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115256

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Celf5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176954.3 NP_795928.2 11 1225
GenBank mRNA AK029796.1 - 11 2399
AK132321.1 - 11 1191
AK158669.1 - 10 1002
BC079845.1 - 11 1196

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001135603, XM_008765105, XM_006240942, XM_008765104, XM_006240941, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0