This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC4B SIRNA Assays ›
Gene Symbol:
Lrrc4b
Gene Name:
leucine rich repeat containing 4B
Gene Aliases:
A830007M12, Lrig4
Chromosome Location:
Chr.7: 44442487 - 44463351 on Build GRCm38
UniGene ID:
Mm.44173
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s114732

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrc4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198250.1 NP_937893.1 3 447
GenBank mRNA AK140418.1 - 2 394
BC053486.1 - 2 311
BC060263.1 - 3 447 5686281

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of synapse assembly
receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0