This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LPCAT2 SIRNA Assays ›
Gene Symbol:
Lpcat2
Gene Name:
lysophosphatidylcholine acyltransferase 2
Gene Aliases:
A330042H22, Aytl1, Aytl1a, lpafat1
Chromosome Location:
Chr.8: 92855330 - 92919279 on Build GRCm38
UniGene ID:
Mm.21463
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s114510

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lpcat2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173014.1 NP_766602.1 10, 11 1162
GenBank mRNA AB244716.1 - 10, 11 1057
AK039431.1 - 10, 11 1162
BC144805.1 - 9, 10 1056
BC144806.1 - 8, 9 997 9053354
BC150666.1 - 8, 9 997 9087577

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
platelet activating factor biosynthetic process
metabolic process
phospholipid biosynthetic process
phosphatidylcholine acyl-chain remodeling
membrane organization
1-acylglycerol-3-phosphate O-acyltransferase activity
calcium ion binding
transferase activity
transferase activity, transferring acyl groups
metal ion binding
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0