This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP512 SIRNA Assays ›
Gene Symbol:
Zfp512
Gene Name:
zinc finger protein 512
Gene Aliases:
2500002M11Rik, AI450173, D230008H22, Znf512
Chromosome Location:
Chr.5: 31452416 - 31481754 on Build GRCm38
UniGene ID:
Mm.259127
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s114388

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp512
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172993.3 NP_766581.2 2 191
GenBank mRNA AK084205.1 - 2 138
AK146353.1 - 2 155
AK149921.1 - 2 138
AK154918.1 - 2 138
AK173267.1 - 2 166
BC066136.1 - 2 114
BC089160.1 - 2 113 5390706

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006239834, XM_006239840, XM_006239836.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header