This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PSME2 SIRNA Assays ›
Gene Symbol:
PSME2
Gene Name:
proteasome activator subunit 2
Gene Aliases:
PA28B, PA28beta, REGbeta
Chromosome Location:
Chr.14: 24143365 - 24147316 on Build GRCh38
UniGene ID:
Hs.434081
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s11427

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PSME2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002818.2 NP_002809.2 7 440
XM_006720213.1 XP_006720276.1 7 1234
GenBank mRNA AK026580.1 - 7 461
AK225876.1 - 7 403
AY771595.1 - 7 389
BC004368.1 - 7 390 3640584
BC019885.1 - 7 390 4994572
BC072025.1 - 7 389 6270762
BX161498.1 - 7 1015
CR541657.1 - 7 375
CR541743.1 - 7 375
D45248.1 - 7 440

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
positive regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
regulation of proteasomal protein catabolic process
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
regulation of G1/S transition of mitotic cell cycle
endopeptidase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0