This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPHN SIRNA Assays ›
Gene Symbol:
Gphn
Gene Name:
gephyrin
Gene Aliases:
5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph
Chromosome Location:
Chr.12: 78224153 - 78684772 on Build GRCm38
UniGene ID:
Mm.341742
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s114051

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gphn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145965.2 NP_666077.2 17 1818
NM_172952.3 NP_766540.2 18 1809
GenBank mRNA AK081716.1 - 1 2796
AK082353.1 - 18 1731
AK144569.1 - 17 1821
BC027112.1 - 6 605
BC127058.1 - 17 1740 40109771

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_022865, X66366, EU735541.

Gene Ontology Categories:

Function(s) Process(es)

Mo-molybdopterin cofactor biosynthetic process
establishment of synaptic specificity at neuromuscular junction
metabolic process
molybdenum incorporation into molybdenum-molybdopterin complex
molybdopterin cofactor biosynthetic process
establishment of protein localization
protein homooligomerization
glycine receptor clustering
nucleotide binding
catalytic activity
receptor binding
protein binding
ATP binding
cytoskeletal protein binding
transferase activity
protein complex scaffold
protein homodimerization activity
metal ion binding
molybdopterin adenylyltransferase activity
molybdopterin molybdotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0