This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLK8 SIRNA Assays ›
Gene Symbol:
Klk8
Gene Name:
kallikrein related-peptidase 8
Gene Aliases:
BSP1, Nrpn, Prss19
Chromosome Location:
Chr.7: 43797552 - 43803822 on Build GRCm38
UniGene ID:
Mm.458023
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s113854

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klk8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008940.2 NP_032966.1 4 1012
GenBank mRNA AB074296.1 - 2 274
BC055895.1 - 4 526 4019947
D30785.1 - 4 1012

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: AJ005641.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
memory
cell death
response to wounding
negative regulation of myelination
keratinocyte proliferation
negative regulation of axon regeneration
neuron projection morphogenesis
regulation of synapse organization
synapse organization
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0