This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1QTNF5 SIRNA Assays ›
Gene Symbol:
C1qtnf5 Mfrp
Gene Name:
C1q and tumor necrosis factor related protein 5
membrane-type frizzled-related protein
Gene Aliases:
Ctrp5
rd6
Chromosome Location:
Chr.9: 44101770 - 44109187 on Build GRCm38
Chromosome Location:
Chr.9: 44101770 - 44109187 on Build GRCm38
UniGene ID:
Mm.227110
Mm.227110
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s113851

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1qtnf5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190313.1 NP_001177242.1 12 1567
NM_145613.4 NP_663588.2 12 1585
GenBank mRNA AF469650.1 - 12 1513
AK053629.1 - 12 1568
Gene Symbol: Mfrp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190314.1 NP_001177243.1 12 1567
NM_147126.3 NP_667337.1 12 1585

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein secretion
inner ear development
protein trimerization
visual perception
eye photoreceptor cell development
retina development in camera-type eye
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header