This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR763 SIRNA Assays ›
Gene Symbol:
Olfr763 Olfr9
Gene Name:
olfactory receptor 763
olfactory receptor 9
Gene Aliases:
MOR269-2
MOR269-3
Chromosome Location:
Chr.10: 129011287 - 129012216 on Build GRCm38
Chromosome Location:
Chr.10: 128989914 - 128990852 on Build GRCm38
UniGene ID:
Mm.377724
Mm.377111
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s113120

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr763
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146862.1 NP_667073.1 1 106
GenBank mRNA BC119229.1 - 1 171 8733978
BC119231.1 - 1 171 8733980
Gene Symbol: Olfr9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146861.1 NP_667072.1 1 106
GenBank mRNA BC116994.1 - 1 171 40126313
BC119365.1 - 1 171 8734114

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000708, NM_001000054.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
signal transduction
response to stimulus
olfactory receptor activity
signal transducer activity
G-protein coupled receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0