This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR1263 SIRNA Assays ›
Gene Symbol:
Olfr1263 Olfr1261
Gene Name:
olfactory receptor 1263
olfactory receptor 1261
Gene Aliases:
MOR234-2
MOR234-3
Chromosome Location:
Chr.2: 90014932 - 90015852 on Build GRCm38
Chromosome Location:
Chr.2: 89993395 - 89994315 on Build GRCm38
UniGene ID:
Mm.223219
Mm.246628
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s112943

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1263
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146794.1 NP_667005.1 1 220
GenBank mRNA BC119193.1 - 1 261 8733942
BC119197.1 - 1 261 8733946
Gene Symbol: Olfr1261
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146474.1 NP_666685.1 1 220
GenBank mRNA AB076075.1 - 1 220
BC119116.1 - 1 252 8733865
BC119144.1 - 1 252 8733893

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000620, NM_001000363, NM_001000619.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0