siRNA Details
Gene Symbol: Olfr255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005780.1 | NP_001005780.1 | 2 | 616 |

Gene Symbol: Olfr255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005780.1 | NP_001005780.1 | 2 | 616 |