This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR1437 SIRNA Assays ›
Gene Symbol:
Olfr1437 Olfr1434 Olfr1436
Gene Name:
olfactory receptor 1437
olfactory receptor 1434
olfactory receptor 1436
Gene Aliases:
MOR214-6
MOR214-4, Olfr1433, Olfr1433-ps1
MOR214-2
Chromosome Location:
Chr.19: 12321887 - 12325066 on Build GRCm38
Chromosome Location:
Chr.19: 12279854 - 12283988 on Build GRCm38
Chromosome Location:
Chr.19: 12298183 - 12299130 on Build GRCm38
UniGene ID:
Mm.432096
Mm.271521
Mm.377636
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s111283

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1437
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011839.1 NP_001011839.1 1 232
Gene Symbol: Olfr1434
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146685.3 NP_666896.2 2 369
GenBank mRNA BC132136.1 - 1 297 40130413
BC138096.1 - 1 297 8861115
BC147691.1 - 1 298 9056664
BC147692.1 - 1 298 9056665
Gene Symbol: Olfr1436
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146687.2 NP_666898.2 1 241
GenBank mRNA BC127991.1 - 1 245 40135681

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001001041.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0