This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL8 SIRNA Assays ›
Gene Symbol:
Klhl8
Gene Name:
kelch-like 8
Gene Aliases:
2310001P09Rik, D5Ertd431e
Chromosome Location:
Chr.5: 103862050 - 103911273 on Build GRCm38
UniGene ID:
Mm.179871
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s110805

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klhl8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178741.3 NP_848856.1 7 1545
GenBank mRNA AK031028.1 - 7 1546
AK034618.1 - 7 1510
AK049960.1 - 6 1433
AY101769.1 - 6 1290
BC086802.1 - 6 1406 30612176

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0