This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FRMPD3 SIRNA Assays ›
Gene Symbol:
Frmpd3
Gene Name:
FERM and PDZ domain containing 3
Gene Aliases:
B930041G04, EG245643
Chromosome Location:
Chr.X: 140293989 - 140394555 on Build GRCm38
UniGene ID:
Mm.208683
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s110648

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Frmpd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177750.6 NP_808418.1 4 494
GenBank mRNA AK047241.1 - 2 2214
BC117734.1 - 2 150 40089821
BC117735.1 - 2 150 40089822
BC129957.1 - 2 150 40089827
BC129958.1 - 2 150 40089829

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003749479.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0