This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ABCC12 SIRNA Assays ›
Gene Symbol:
Abcc12
Gene Name:
ATP-binding cassette, sub-family C (CFTR/MRP), member 12
Gene Aliases:
4930467B22Rik, MRP9
Chromosome Location:
Chr.8: 86504576 - 86580690 on Build GRCm38
UniGene ID:
Mm.67485
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s110336

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Abcc12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172912.4 NP_766500.3 28 3941
GenBank mRNA AF502146.1 - 28 3941
AF514414.1 - 29 4074
AF514415.1 - 29 4041
AK029684.1 - 14 1756
AK133175.1 - 4117
BC138380.1 - 28 3934 8861400
BC138381.1 - 28 3934 8861401
BC171952.1 - 28 3934 9054238

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_199377, XM_008772375, XM_008772377, XM_006255241, XM_008772376, and others.

Gene Ontology Categories:

Function(s) Process(es)

transport
transmembrane transport
nucleotide binding
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0