This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC13 SIRNA Assays ›
Gene Symbol:
Zdhhc13
Gene Name:
zinc finger, DHHC domain containing 13
Gene Aliases:
2410004E01Rik, C530010M18, HIP3RP, Hip14l, kojak, skc4
Chromosome Location:
Chr.7: 48789003 - 48827440 on Build GRCm38
UniGene ID:
Mm.279116
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s110198

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028031.3 NP_082307.1 15 1637
GenBank mRNA AK010382.1 - 15 1637
AK049640.1 - 14 1466
AK145855.1 - 15 1557
AK146196.1 - 15 1609
BC019493.1 - 15 1556
BC046599.1 - 15 1556 6311721

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
signal transduction
magnesium ion transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
cation transmembrane transport
signal transducer activity
zinc ion binding
magnesium ion transmembrane transporter activity
palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
protein-cysteine S-palmitoyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0