This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700024P16RIK SIRNA Assays ›
Gene Symbol:
1700024P16Rik
Gene Name:
RIKEN cDNA 1700024P16 gene
Gene Aliases:
C1orf168, Gm620
Chromosome Location:
Chr.4: 104833834 - 105016864 on Build GRCm38
UniGene ID:
Mm.65181
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s109793

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700024P16Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162980.1 NP_001156452.1 18 2360
NM_001285952.1 NP_001272881.1 19 2438
NM_001285953.1 NP_001272882.1 18 2293
GenBank mRNA AK028077.1 - 18 2300
AK133762.1 - 18 2231
BC137723.1 - 18 2360 8860742
BC144857.1 - 19 2437 9053405
BC144858.1 - 19 2401 9053406

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008763964.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0