This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP2R2C SIRNA Assays ›
Gene Symbol:
PPP2R2C
Gene Name:
protein phosphatase 2 regulatory subunit Bgamma
Gene Aliases:
B55-GAMMA, IMYPNO, IMYPNO1, PR52, PR55G
Chromosome Location:
Chr.4: 6320578 - 6563600 on Build GRCh38
UniGene ID:
Hs.479069
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s10973

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PPP2R2C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206994.1 NP_001193923.1 8 982
NM_001206995.1 NP_001193924.1 8 1086
NM_001206996.1 NP_001193925.1 8 1136
NM_020416.3 NP_065149.2 7 1274
NM_181876.2 NP_870991.1 7 928
XM_005247979.4 XP_005248036.1 3 1119
XM_011513495.1 XP_011511797.1 7 1023
XM_011513496.1 XP_011511798.1 7 1038
GenBank mRNA AF086924.1 - 7 958
AF145027.1 - 7 1121
AK093115.1 - 7 928
AK122849.1 - 3 2935
AK296474.1 - 8 982
AK316003.1 - 9 1096
BC032954.2 - 8 1136 4823280

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphatase type 2A activity
protein binding
protein phosphatase type 2A regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0