This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADAMTS4 SIRNA Assays ›
Gene Symbol:
Adamts4
Gene Name:
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4
Gene Aliases:
3830423K05, ADAM-TS4, ADAMTS-2, ADMP-1, mKIAA0688
Chromosome Location:
Chr.1: 171250422 - 171259922 on Build GRCm38
UniGene ID:
Mm.491147
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s109409

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adamts4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172845.2 NP_766433.1 4 1553
GenBank mRNA AF449483.1 - 1 46
AK028364.1 - 1554
AK076295.1 - 3 928
AK083534.1 - 4 1553
AK156429.1 - 2 136
AK157287.1 - 1554
AK164900.1 - 4 1549
AK173001.1 - 1431
BC027773.1 - 4 1187 5345809

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
defense response to bacterium
protease binding
metalloendopeptidase activity
protein binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0