This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLCO5A1 SIRNA Assays ›
Gene Symbol:
Slco5a1
Gene Name:
solute carrier organic anion transporter family, member 5A1
Gene Aliases:
4732454B15, A630033C23Rik, AA407151
Chromosome Location:
Chr.1: 12866549 - 12992932 on Build GRCm38
UniGene ID:
Mm.24481
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s109346

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slco5a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172841.2 NP_766429.2 8 2716
GenBank mRNA AV282837.2 - 2 85

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001146009, NM_030958, BC137424, BC144618, BC139755, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0