This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC35G1 SIRNA Assays ›
Gene Symbol:
Slc35g1
Gene Name:
solute carrier family 35, member G1
Gene Aliases:
AA590464, D330039I19Rik, Tmem20
Chromosome Location:
Chr.19: 38395980 - 38405607 on Build GRCm38
UniGene ID:
Mm.262085
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s109319

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc35g1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175507.3 NP_780716.1 3 859
GenBank mRNA AK041618.1 - 3 859
AK052365.1 - 3 763
BC137663.1 - 3 870 8860682
BC137664.1 - 3 870 8860683

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cytosolic calcium ion concentration
calcium ion export from cell
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0