This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RIMBP3 SIRNA Assays ›
Gene Symbol:
Rimbp3
Gene Name:
RIMS binding protein 3
Gene Aliases:
Gm1759, Gm603, RIM-BP3, mKIAA1666
Chromosome Location:
Chr.16: 17208135 - 17213982 on Build GRCm38
UniGene ID:
Mm.86671
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s109077

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rimbp3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033338.3 NP_001028510.2 1 3229
GenBank mRNA AK133200.1 - 1 2380
AK220410.1 - 1 1249
BC141386.1 - 1 2373 9055999

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_221302.

Gene Ontology Categories:

Function(s) Process(es)

spermatid development
fertilization
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0