This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POU2F1 SIRNA Assays ›
Gene Symbol:
POU2F1
Gene Name:
POU class 2 homeobox 1
Gene Aliases:
OCT1, OTF1, oct-1B
Chromosome Location:
Chr.1: 167220829 - 167427345 on Build GRCh38
UniGene ID:
Hs.283402
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s10847

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: POU2F1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198783.1 NP_001185712.1 8 941
NM_001198786.1 NP_001185715.1 7 619
NM_002697.3 NP_002688.3 8 808
NR_037163.1 9 852
XM_011509653.2 XP_011507955.1 9 760
XM_011509654.2 XP_011507956.1 8 721
XM_011509655.1 XP_011507957.1 9 760
XM_017001508.1 XP_016856997.1 9 760
GenBank mRNA AK091438.1 - 10 1236
AK302525.1 - 9 847
AK303201.1 - 7 557
AK308004.1 - 9 847
AY113189.1 - 8 760
BC001664.2 - 8 724 2966289
BC003571.1 - 8 724 3544378
BC007388.2 - 4 394
BC052274.1 - 9 832 4622256
KT438684.1 - 10 919

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

snRNA transcription from RNA polymerase II promoter
negative regulation of transcription, DNA-templated
liver regeneration
transcription regulatory region sequence-specific DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
receptor binding
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0