This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM205 SIRNA Assays ›
Gene Symbol:
Tmem205
Gene Name:
transmembrane protein 205
Gene Aliases:
-
Chromosome Location:
Chr.9: 21921009 - 21927535 on Build GRCm38
UniGene ID:
Mm.253329
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s108031

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem205
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253867.1 NP_001240796.1 2, 3 293
NM_001253868.1 NP_001240797.1 2, 3 366
NM_001253869.1 NP_001240798.1 2, 3 225
NM_001253870.1 NP_001240799.1 2, 3 353
NM_178577.5 NP_848692.2 2, 3 361
GenBank mRNA AI573565.1 - 2, 3 177
AK161954.1 - 1 1411
BC010787.1 - 2, 3 288 4211629
CA326029.1 - 2, 3 225
CN840781.1 - 2, 3 281

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0