This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTC13 SIRNA Assays ›
Gene Symbol:
Ttc13
Gene Name:
tetratricopeptide repeat domain 13
Gene Aliases:
BC017545
Chromosome Location:
Chr.8: 124671327 - 124722013 on Build GRCm38
UniGene ID:
Mm.38436
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s107987

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttc13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145607.3 NP_663582.2 2 271
GenBank mRNA AK051200.1 - 2 278
AK144842.1 - 2 272
AK148102.1 - 2 270
AK160894.1 - 2 280
AK212172.1 - 2 197
BC072577.1 - 2 264
BC079851.1 - 2 260 6401930

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0